Gene Transcripts

Gene Symbol
HNRNPA2B1
Entrez Gene ID
3181
Gene Name
heterogeneous nuclear ribonucleoprotein A2/B1
Gene Aliases
HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, IBMPFD2, RNPA2, SNRPB1
Chromosome Location
Chr. 7: 26189927 - 26200793 on Build GRCh38
UniGene ID
Hs.487774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002137.3 NP_002128.1 6 852
NM_031243.2 NP_112533.1 7 888
XM_005249729.1 XP_005249786.1 7 870
XM_017012109.1 XP_016867598.1 6 836
XM_017012110.1 XP_016867599.1 6 836
GenBank mRNA AK129839.1 - 6 806
AK289429.1 - 7 841
BX537494.1 - 5 3685
BX641084.1 - 5 4426
M29064.1 - 7 821
M29065.1 - 6 791

View Full Product Details