Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000090.3 | NP_000081.1 | 2 | 270 | |
GenBank mRNA | AK308010.1 | - | 2 | 270 | |
BC028178.1 | - | 2 | 268 | 5405119 | |
BP374999.1 | - | 2 | 270 | ||
X07240.1 | - | 2 | 153 | ||
X14420.1 | - | 2 | 255 |