Gene Transcripts

Gene Symbol
CALB2
Entrez Gene ID
794
Gene Name
calbindin 2
Gene Aliases
CAB29, CAL2, CR
Chromosome Location
Chr. 16: 71358713 - 71390438 on Build GRCh38
UniGene ID
Hs.106857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001740.4 NP_001731.2 11 1159
NM_007088.3 NP_009019.1 9 1065
NR_027910.2 10 1119
GenBank mRNA AK222495.1 - 11 1149
AK222648.1 - 11 1149
AK291096.1 - 11 1149
BC015484.2 - 11 1134 3847342
X56667.1 - 11 1122

View Full Product Details