Gene Transcripts

Gene Symbol
CSNK2A1
Entrez Gene ID
1457
Gene Name
casein kinase 2 alpha 1
Gene Aliases
CK2A1, CKII, CSNK2A3
Chromosome Location
Chr. 20: 482694 - 543838 on Build GRCh38
UniGene ID
Hs.644056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001895.3 NP_001886.1 13 2580
NM_177559.2 NP_808227.1 14 2697
NM_177560.2 NP_808228.1 12 2370
GenBank mRNA BC050036.1 - 13 2469
BC053532.1 - 12 2399 3908058
BC071167.1 - 12 2311 5772615

View Full Product Details