Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004429.4 | NP_004420.1 | 2 | 932 | |
GenBank mRNA | BC016649.1 | - | 2 | 932 | |
BC052979.1 | - | 2 | 951 | 6164440 | |
L37361.1 | - | 2 | 678 | ||
U09303.1 | - | 2 | 853 | ||
U09304.1 | - | 2 | 459 |