Gene Transcripts

Gene Symbol
CRHR1
Entrez Gene ID
1394
Gene Name
corticotropin releasing hormone receptor 1
Gene Aliases
CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L
Chromosome Location
Chr. 17: 45784280 - 45835828 on Build GRCh38
UniGene ID
Hs.417628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145146.1 NP_001138618.1 5 608
NM_001145147.1 NP_001138619.1 4 488
NM_001145148.1 NP_001138620.1 5 608
NM_001303018.1 NP_001289947.1 3 402
NM_001303020.1 NP_001289949.1 4 522
NM_004382.4 NP_004373.2 5 608
GenBank mRNA AB451466.1 - 5 343
AF180301.1 - 5 343
AF369651.1 - 2 40
AF374231.1 - 5 356
AY429529.1 - 3 199
AY457172.1 - 5 343
BC096836.1 - 5 363 6971902
EU012435.1 - 4 257
EU176016.1 - 2 40
FJ200483.1 - 3 137
HM852949.1 - 5 578
L23332.1 - 5 380
L23333.1 - 5 343
U16273.1 - 4 241
X72304.1 - 5 569

Gene Symbol
MGC57346-CRHR1
Entrez Gene ID
104909134
Gene Name
MGC57346-CRHR1 readthrough
Gene Aliases
Chromosome Location
Chr. 17: 45620329 - 45835828 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256299.2 NP_001243228.1 7 743
NM_001303016.1 NP_001289945.1 6 689
GenBank mRNA AK295559.1 - 6 715
AK297590.1 - 6 671
FJ543100.1 - 7 728

View Full Product Details