Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004952.4 | NP_004943.1 | 2 | 246 | |
GenBank mRNA | AK296030.1 | - | 2 | 230 | |
AK316248.1 | - | 2 | 209 | ||
BC017722.1 | - | 2 | 246 | 4397263 | |
BC110406.1 | - | 2 | 236 | 5732044 | |
L37360.1 | - | 2 | 194 | ||
U14187.1 | - | 2 | 233 |