Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
1411
Gene Name
crystallin beta A1
Gene Aliases
CRYB1, CTRCT10
Chromosome Location
Chr. 17: 29246857 - 29254494 on Build GRCh38
UniGene ID
Hs.46275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005208.4 NP_005199.2 6 706
XM_017024198.1 XP_016879687.1 7 1002
GenBank mRNA BC113693.1 - 6 700

View Full Product Details