Gene Transcripts

Gene Symbol
ACPP
Entrez Gene ID
55
Gene Name
acid phosphatase, prostate
Gene Aliases
5'-NT, ACP-3, ACP3
Chromosome Location
Chr. 3: 132317367 - 132368765 on Build GRCh38
UniGene ID
Hs.433060
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099.4 NP_001090.2 10 1404
NM_001292037.1 NP_001278966.1 9 1305
GenBank mRNA BC008493.1 - 10 1361
BC016344.1 - 10 1368 3950907
M34840.1 - 10 1329
X52174.1 - 10 1338
X53605.1 - 10 1351

View Full Product Details