Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134.2 | NP_001125.1 | 11, 12 | 1474 | |
GenBank mRNA | AK297669.1 | - | 11, 12 | 1386 | |
AK297674.1 | - | 11, 12 | 1327 | ||
AK314817.1 | - | 11, 12 | 1468 | ||
BC027881.1 | - | 11, 12 | 1460 | 5208299 | |
V01514.1 | - | 11, 12 | 1474 |