Gene Transcripts

Gene Symbol
C3AR1
Entrez Gene ID
719
Gene Name
complement component 3a receptor 1
Gene Aliases
AZ3B, C3AR, HNFAG09
Chromosome Location
Chr. 12: 8058323 - 8066359 on Build GRCh38
UniGene ID
Hs.504656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001326475.1 NP_001313404.1 2 368
NM_001326477.1 NP_001313406.1 2 428
NM_004054.3 NP_004045.1 2 371
GenBank mRNA AK223388.1 - 2 281
AK290242.1 - 2 259
AK312458.1 - 2 259
BC020742.1 - 2 259 4690283
DA351375.1 - 2 372
DB013722.1 - 2 294
DB274838.1 - 2 256
U28488.1 - 2 248
U62027.1 - 2 319
Z73157.1 - 1 167

View Full Product Details