Gene Transcripts

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
Chromosome Location
Chr. 15: 74541177 - 74598131 on Build GRCh38
UniGene ID
Hs.647642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307939.1 NP_001294868.1 4 911
NM_006465.3 NP_006456.1 4 911
GenBank mRNA AB208830.1 - 4 830
AF116846.1 - 4 843
AK298716.1 - 4 836
AK302034.1 - 4 911
BC041792.1 - 4 839 5297774
BC045545.1 - 4 885
BC060824.1 - 4 824

View Full Product Details