Gene Transcripts

Gene Symbol
SLC9A8
Entrez Gene ID
23315
Gene Name
solute carrier family 9 member A8
Gene Aliases
NHE-8, NHE8
Chromosome Location
Chr. 20: 49812713 - 49892242 on Build GRCh38
UniGene ID
Hs.444202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001260491.1 NP_001247420.1 16 4409
NM_015266.2 NP_056081.1 16 4361
NR_048537.1 15 4326
NR_048538.1 14 4242
NR_048539.1 14 4198
NR_048540.1 12 4024
XM_006723756.1 XP_006723819.1 17 4322
XM_011528736.1 XP_011527038.1 17 4370
XM_011528737.1 XP_011527039.1 16 4243
XM_011528738.1 XP_011527040.1 16 4253
XM_011528739.1 XP_011527041.1 16 4247
XM_011528741.1 XP_011527043.1 15 4154
XM_011528742.1 XP_011527044.1 15 4143
XM_011528743.2 XP_011527045.1 9 3507
XM_011528744.2 XP_011527046.1 8 3338
XM_017027755.1 XP_016883244.1 16 4231
XM_017027756.1 XP_016883245.1 17 4319
GenBank mRNA AB023156.2 - 16 4193
AL049469.1 - 1 430
AL833574.1 - 5 3060

View Full Product Details