Gene Transcripts

Gene Symbol
SRPRB
Entrez Gene ID
58477
Gene Name
SRP receptor beta subunit
Gene Aliases
APMCF1
Chromosome Location
Chr. 3: 133784033 - 133821492 on Build GRCh38
UniGene ID
Hs.12152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021203.3 NP_067026.3 5 683
GenBank mRNA AF086914.1 - 4 421
AF141882.3 - 4 414
AK027307.1 - 1 926
AK027525.1 - 4 414
AK075531.1 - 4 452
BC063001.1 - 4 426 5185997
BC065299.1 - 4 398 6146939

View Full Product Details