Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282405.1 | NP_001269334.1 | 3 | 772 | |
NM_030759.4 | NP_110386.2 | 4 | 857 | ||
XM_006717810.3 | XP_006717873.1 | 5 | 1042 | ||
GenBank mRNA | AF267866.1 | - | 4 | 808 | |
AK054957.1 | - | 4 | 815 | ||
AK225256.1 | - | 4 | 766 | ||
AK225257.1 | - | 4 | 768 | ||
AK301656.1 | - | 3 | 694 | ||
AL136656.1 | - | 4 | 812 | ||
AY267839.1 | - | 5 | 595 | ||
AY267840.1 | - | 4 | 745 | ||
BC001345.1 | - | 1 | 441 | ||
BC011707.1 | - | 4 | 731 | 3687848 |