Gene Transcripts

Gene Symbol
MTG1
Entrez Gene ID
92170
Gene Name
mitochondrial ribosome associated GTPase 1
Gene Aliases
GTP, GTPBP7
Chromosome Location
Chr. 10: 133394117 - 133420670 on Build GRCh38
UniGene ID
Hs.501578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138384.2 NP_612393.2 11 1416
GenBank mRNA AK074976.1 - 10 1895
AK095872.1 - 5 2285
AK125012.1 - 11 2261
BC000920.2 - 9 1125
BC004409.1 - 11 1375
BC026039.1 - 11 1388 5444745
BC026725.1 - 11 1295
BC035721.1 - 11 1320 5588933

View Full Product Details