Gene Transcripts

Gene Symbol
RASGEF1A
Entrez Gene ID
221002
Gene Name
RasGEF domain family member 1A
Gene Aliases
CG4853
Chromosome Location
Chr. 10: 43194533 - 43266919 on Build GRCh38
UniGene ID
Hs.125293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282862.1 NP_001269791.1 13 1593
NM_145313.3 NP_660356.2 13 1565
XM_005271809.3 XP_005271866.1 12 1443
XM_011539500.2 XP_011537802.1 12 1553
GenBank mRNA AK095136.1 - 13 1593
AK127432.1 - 13 3991
BC022548.1 - 13 1565 4814750

View Full Product Details