Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004427.3 | NP_004418.2 | 7 | 1980 | |
NM_198040.2 | NP_932157.1 | 14 | 3287 | ||
XM_005270570.1 | XP_005270627.1 | 7 | 1916 | ||
XM_017000513.1 | XP_016856002.1 | 15 | 3786 | ||
XM_017000514.1 | XP_016856003.1 | 15 | 3398 | ||
XM_017000515.1 | XP_016856004.1 | 15 | 3491 | ||
XM_017000516.1 | XP_016856005.1 | 15 | 3720 | ||
XM_017000517.1 | XP_016856006.1 | 15 | 3332 | ||
XM_017000518.1 | XP_016856007.1 | 15 | 3425 | ||
XM_017000519.1 | XP_016856008.1 | 16 | 3875 | ||
XM_017000520.1 | XP_016856009.1 | 14 | 3245 | ||
XM_017000521.1 | XP_016856010.1 | 14 | 3664 | ||
GenBank mRNA | AJ419231.1 | - | 14 | 3287 | |
AK056429.1 | - | 8 | 2217 | ||
AK091856.1 | - | 1 | 1716 | ||
AK128821.1 | - | 8 | 1965 | ||
BC015450.1 | - | 3 | 1127 | ||
BC018602.1 | - | 7 | 1986 | ||
BC028396.1 | - | 7 | 1951 | ||
BC029269.1 | - | 7 | 1822 | ||
BC068573.1 | - | 7 | 1874 | ||
BC092492.1 | - | 7 | 1874 | ||
BC110863.1 | - | 7 | 1873 | ||
BC130630.1 | - | 13 | 3203 | 40146661 | |
BC144121.1 | - | 13 | 3203 | 9052641 | |
BX537498.1 | - | 1 | 1952 | ||
U89278.1 | - | 7 | 1961 |