Gene Transcripts

Gene Symbol
C1QC
Entrez Gene ID
714
Gene Name
complement C1q C chain
Gene Aliases
C1Q-C, C1QG
Chromosome Location
Chr. 1: 22643625 - 22648110 on Build GRCh38
UniGene ID
Hs.467753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114101.1 NP_001107573.1 3 721
NM_172369.3 NP_758957.2 3 698
GenBank mRNA AF087892.1 - 3 696
AK057792.1 - 3 713
AK130613.1 - 3 696
AK226152.1 - 1 3993
BC009016.1 - 3 663 4212772

View Full Product Details