Gene Transcripts
- Gene Symbol
- C9orf84
- Entrez Gene ID
- 158401
- Gene Name
- chromosome 9 open reading frame 84
- Gene Aliases
- Chromosome Location
- Chr. 9: 111686175 - 111795949 on Build GRCh38
- UniGene ID
- Hs.428209
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109816.1 |
|
3, 4 |
317 |
|
XM_006716974.2 |
XP_006717037.1
|
3, 4 |
301 |
|
XM_006716975.2 |
XP_006717038.1
|
3, 4 |
301 |
|
XM_011518309.1 |
XP_011516611.1
|
3, 4 |
301 |
|
XM_017014340.1 |
XP_016869829.1
|
3, 4 |
1258 |
|
XM_017014341.1 |
XP_016869830.1
|
3, 4 |
301 |
|
GenBank mRNA |
AK057341.1 |
- |
3, 4 |
252 |
|
AL704322.1 |
- |
3, 4 |
318 |
|
BC112357.1 |
- |
3, 4 |
245 |
|
BC144680.1 |
- |
3, 4 |
241 |
|
View Full Product Details