Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_203301.3 | NP_976046.1 | 4 | 2250 | |
GenBank mRNA | AK294140.1 | - | 2 | 1094 | |
BC042535.1 | - | 2 | 729 | ||
BC053537.1 | - | 4 | 2167 | 5181903 | |
BC068566.1 | - | 4 | 2222 | 5270962 |