Gene Transcripts

Gene Symbol
MMRN1
Entrez Gene ID
22915
Gene Name
multimerin 1
Gene Aliases
ECM, EMILIN4, GPIa*, MMRN
Chromosome Location
Chr. 4: 89879539 - 89954629 on Build GRCh38
UniGene ID
Hs.268107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007351.2 NP_031377.2 5 1137
XM_005262856.2 XP_005262913.1 4 1057
XM_017007893.1 XP_016863382.1 6 1378
GenBank mRNA AK125557.1 - 6 1084
AK302421.1 - 5 516
AK313566.1 - 5 1162
BC063848.1 - 5 1137 6180674
U27109.1 - 5 1137

View Full Product Details