Gene Transcripts

Gene Symbol
ZBTB11
Entrez Gene ID
27107
Gene Name
zinc finger and BTB domain containing 11
Gene Aliases
ZNF-U69274, ZNF913
Chromosome Location
Chr. 3: 101649439 - 101677144 on Build GRCh38
UniGene ID
Hs.655286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014415.3 NP_055230.2 1 513
GenBank mRNA AK312859.1 - 1 505
BC111700.1 - 1 420 40038346
DA907277.1 - 1 513
U69274.1 - 1 444

View Full Product Details