Gene Transcripts

Gene Symbol
EXOSC4
Entrez Gene ID
54512
Gene Name
exosome component 4
Gene Aliases
RRP41, RRP41A, Rrp41p, SKI6, Ski6p, hRrp41p, p12A
Chromosome Location
Chr. 8: 144064092 - 144080648 on Build GRCh38
UniGene ID
Hs.632041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019037.2 NP_061910.1 3 519
XM_011517134.2 XP_011515436.1 3 377
GenBank mRNA AF281133.1 - 3 512
AK000598.1 - 3 436
BC002777.2 - 3 433 3613853
BM839994.1 - 3 519

View Full Product Details