Gene Transcripts

Gene Symbol
RBMXL1
Entrez Gene ID
494115
Gene Name
RNA binding motif protein, X-linked like 1
Gene Aliases
RBM1
Chromosome Location
Chr. 1: 88979456 - 88992960 on Build GRCh38
UniGene ID
Hs.481898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162536.2 NP_001156008.1 3 2896
NM_019610.5 NP_062556.2 2 2796
GenBank mRNA AL832554.1 - 2 2649
AL832785.1 - 1 1740
BC012942.1 - 2 2455 3875565

Gene Symbol
RBMX
Entrez Gene ID
27316
Gene Name
RNA binding motif protein, X-linked
Gene Aliases
HNRNPG, HNRPG, MRXS11, RBMXP1, RBMXRT, RNMX, hnRNP-G
Chromosome Location
Chr. X: 136869194 - 136880780 on Build GRCh38
UniGene ID
Hs.380118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164803.1 NP_001158275.1 8 847
GenBank mRNA AK097941.1 - 1 548
BX647131.1 - 8 3680

View Full Product Details