Gene Transcripts

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
Chromosome Location
Chr. 8: 12945674 - 13029777 on Build GRCh38
UniGene ID
Hs.202521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB040889.1 - 2 718
AK307825.1 - 2 969
BC016633.2 - 2 297
DR001645.1 - 2 425

View Full Product Details