Gene Transcripts
- Gene Symbol
- ZFAT
- Entrez Gene ID
- 57623
- Gene Name
- zinc finger and AT-hook domain containing
- Gene Aliases
- AITD3, ZFAT1, ZNF406
- Chromosome Location
- Chr. 8: 134477788 - 134713049 on Build GRCh38
- UniGene ID
- Hs.446172
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001029939.3 |
NP_001025110.2
|
4, 5 |
715 |
|
NM_001167583.2 |
NP_001161055.1
|
3, 4 |
556 |
|
NM_001174158.1 |
NP_001167629.1
|
3, 4 |
556 |
|
NM_001289394.1 |
NP_001276323.1
|
4, 5 |
792 |
|
NM_020863.3 |
NP_065914.2
|
3, 4 |
633 |
|
NR_110323.1 |
|
3, 4 |
633 |
|
XM_011517203.1 |
XP_011515505.1
|
3, 4 |
530 |
|
XM_011517204.2 |
XP_011515506.1
|
2, 3 |
353 |
|
XM_011517206.1 |
XP_011515508.1
|
3, 4 |
554 |
|
XM_017013716.1 |
XP_016869205.1
|
2, 3 |
428 |
|
GenBank mRNA |
AB040918.1 |
- |
1, 2 |
31 |
|
AB167738.1 |
- |
3, 4 |
622 |
|
AB167739.1 |
- |
4, 5 |
781 |
|
AB167740.1 |
- |
4, 5 |
708 |
|
AB167741.1 |
- |
3, 4 |
549 |
|
AK300133.1 |
- |
3, 4 |
545 |
|
AK300324.1 |
- |
3, 4 |
552 |
|
AK300471.1 |
- |
3, 4 |
618 |
|
AK311494.1 |
- |
3, 4 |
554 |
|
AL832066.1 |
- |
3, 4 |
633 |
|
BC012989.2 |
- |
3, 4 |
547 |
|
BC025423.2 |
- |
1, 2 |
225 |
|
BC098405.1 |
- |
3, 4 |
556 |
|
BC101766.1 |
- |
3, 4 |
546 |
8069272 |
BC101768.1 |
- |
3, 4 |
546 |
8069274 |
View Full Product Details