Gene Transcripts

Gene Symbol
ATP13A4
Entrez Gene ID
84239
Gene Name
ATPase 13A4
Gene Aliases
Chromosome Location
Chr. 3: 193398967 - 193581725 on Build GRCh38
UniGene ID
Hs.674423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032279.3 NP_115655.2 2 416
XM_011513232.2 XP_011511534.1 2 163
XM_017007318.1 XP_016862807.1 2 52
XM_017007319.1 XP_016862808.1 2 163
GenBank mRNA AK095277.1 - 2 201
AY823162.1 - 2 93
BC101496.1 - 2 187 8069002
DA117883.1 - 2 416

View Full Product Details