Gene Transcripts
- Gene Symbol
- CLNK
- Entrez Gene ID
- 116449
- Gene Name
- cytokine dependent hematopoietic cell linker
- Gene Aliases
- MIST
- Chromosome Location
- Chr. 4: 10489899 - 10734853 on Build GRCh38
- UniGene ID
- Hs.678910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB032369.1 |
- |
9, 10 |
545 |
|
AB110420.1 |
- |
11, 12 |
839 |
|
View Full Product Details