Gene Transcripts

Gene Symbol
DEPDC4
Entrez Gene ID
120863
Gene Name
DEP domain containing 4
Gene Aliases
DEP.4
Chromosome Location
Chr. 12: 100203669 - 100268094 on Build GRCh38
UniGene ID
Hs.653118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319311.1 NP_001306240.1 11 1693
NR_135073.1 8 1348
XM_017018781.1 XP_016874270.1 10 1654
GenBank mRNA AK090824.1 - 8 1348

View Full Product Details