Gene Transcripts

Gene Symbol
FAM27C
Entrez Gene ID
100132948
Gene Name
family with sequence similarity 27 member C
Gene Aliases
FAM27A, FAM27A1, FAM27A3, bA7G23.5
Chromosome Location
Chr. 9: 61854084 - 61855340 on Build GRCh38
UniGene ID
Hs.645763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027421.1 2 429
GenBank mRNA BC002886.1 - 2 360

Gene Symbol
FAM27B
Entrez Gene ID
100133121
Gene Name
family with sequence similarity 27 member B
Gene Aliases
FAM27A2
Chromosome Location
Chr. 9: 67725483 - 67726743 on Build GRCh38
UniGene ID
Hs.744835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027422.1 2 429
GenBank mRNA AK131029.1 - 2 312

Gene Symbol
FAM27E5
Entrez Gene ID
284123
Gene Name
family with sequence similarity E5
Gene Aliases
FAM27L
Chromosome Location
Chr. 17: 22298764 - 22299893 on Build GRCh38
UniGene ID
Hs.348578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028336.1 2 341
GenBank mRNA BC031617.1 - 2 341

View Full Product Details