Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256126.1 | NP_001243055.1 | 9 | 1123 | |
NM_203463.2 | NP_982288.1 | 9 | 1123 | ||
XM_005246440.4 | XP_005246497.1 | 6 | 2414 | ||
XM_017003749.1 | XP_016859238.1 | 6 | 1274 | ||
GenBank mRNA | AK172775.1 | - | 9 | 1123 | |
BC030800.2 | - | 4 | 354 | ||
BC109284.1 | - | 9 | 973 | 40031838 | |
BC109285.1 | - | 9 | 973 | 40031840 | |
BG206657.1 | - | 6 | 529 |