Gene Transcripts

Gene Symbol
C19orf70
Entrez Gene ID
125988
Gene Name
chromosome 19 open reading frame 70
Gene Aliases
MIC13, P117, QIL1
Chromosome Location
Chr. 19: 5678414 - 5680896 on Build GRCh38
UniGene ID
Hs.356626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308240.1 NP_001295169.1 4, 5 873
NM_205767.2 NP_991330.1 3, 4 677
XM_017026248.1 XP_016881737.1 3, 4 618
GenBank mRNA AA292888.1 - 3, 4 444
AJ707830.1 - 3, 4 423
BC009557.1 - 3, 4 311 3899541
BC042386.1 - 3, 4 286 5288954
BC052640.1 - 3, 4 288
BI827162.1 - 3, 4 676

View Full Product Details