Gene Transcripts

Gene Symbol
GFRAL
Entrez Gene ID
389400
Gene Name
GDNF family receptor alpha like
Gene Aliases
C6orf144, GRAL, UNQ9356, bA360D14.1
Chromosome Location
Chr. 6: 55327469 - 55402493 on Build GRCh38
UniGene ID
Hs.526967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207410.2 NP_997293.2 5 599
GenBank mRNA AY358198.1 - 5 600

View Full Product Details