Gene Transcripts

Gene Symbol
PADI6
Entrez Gene ID
353238
Gene Name
peptidyl arginine deiminase 6
Gene Aliases
hPADVI
Chromosome Location
Chr. 1: 17372196 - 17401699 on Build GRCh38
UniGene ID
Hs.531598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207421.4 NP_997304.3 14 1729
GenBank mRNA AY422079.1 - 14 1679
AY443100.1 - 14 1730

View Full Product Details