Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207437.3 | NP_997320.2 | 8 | 1007 | |
XM_005253555.3 | XP_005253612.1 | 8 | 1499 | ||
XM_011538014.2 | XP_011536316.1 | 8 | 1499 | ||
XM_011538015.2 | XP_011536317.1 | 8 | 1499 | ||
XM_011538016.2 | XP_011536318.1 | 8 | 1499 | ||
XM_011538017.2 | XP_011536319.1 | 8 | 1499 | ||
XM_011538020.2 | XP_011536322.1 | 8 | 1507 | ||
XM_017018960.1 | XP_016874449.1 | 8 | 1499 | ||
XM_017018961.1 | XP_016874450.1 | 8 | 1499 | ||
GenBank mRNA | AK125475.1 | - | 3 | 521 | |
BC144575.1 | - | 3 | 497 | ||
BC150622.1 | - | 3 | 497 |