Gene Transcripts

Gene Symbol
FSIP2
Entrez Gene ID
401024
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
Chromosome Location
Chr. 2: 185738895 - 185833289 on Build GRCh38
UniGene ID
Hs.98025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173651.3 NP_775922.3 17 16999
GenBank mRNA AK126051.1 - 1 505
BX647691.1 - 1 224
BX648733.1 - 1 1272

View Full Product Details