Gene Transcripts

Gene Symbol
SAMD12
Entrez Gene ID
401474
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
Chromosome Location
Chr. 8: 118131828 - 118621995 on Build GRCh38
UniGene ID
Hs.359393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207506.2 NP_997389.2 4 1404
XM_017013409.1 XP_016868898.1 4 1416
GenBank mRNA AK096777.1 - 4 1340

View Full Product Details