Gene Transcripts

Gene Symbol
Plxna2
Entrez Gene ID
18845
Gene Name
plexin A2
Gene Aliases
2810428A13Rik, AA589422, AW457381, OCT, PlexA2, Plxn2, mKIAA0463
Chromosome Location
Chr. 1: 194619829 - 194816869 on Build GRCm38
UniGene ID
Mm.2251
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008882.2 NP_032908.2 2 1640
GenBank mRNA BC068155.1 - 2 1422
D86949.1 - 2 1546

View Full Product Details