Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008997.3 | NP_033023.1 | 5 | 769 | |
GenBank mRNA | AB232606.1 | - | 5 | 545 | |
AK134125.1 | - | 5 | 514 | ||
AK146452.1 | - | 5 | 630 | ||
AK146460.1 | - | 5 | 581 | ||
AK146623.1 | - | 5 | 581 | ||
AK146978.1 | - | 5 | 561 | ||
AK147077.1 | - | 5 | 600 | ||
AK148022.1 | - | 5 | 642 | ||
AK148106.1 | - | 5 | 568 | ||
AK188721.1 | - | 3 | 218 | ||
AK203890.1 | - | 3 | 243 | ||
AK205000.1 | - | 3 | 243 | ||
BC054753.1 | - | 5 | 585 | 6411651 | |
BC085270.1 | - | 5 | 578 | 6478784 | |
BC157916.1 | - | 5 | 598 | 9007344 | |
KJ650201.1 | - | 5 | 545 | ||
L26528.1 | - | 5 | 580 |