Gene Transcripts

Gene Symbol
VNN1
Entrez Gene ID
8876
Gene Name
vanin 1
Gene Aliases
HDLCQ8, Tiff66
Chromosome Location
Chr. 6: 132680858 - 132714055 on Build GRCh38
UniGene ID
Hs.12114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004666.2 NP_004657.2 7 2101
GenBank mRNA AJ132099.1 - 7 2095
AK095461.1 - 4 1822
U39664.1 - 7 2096

View Full Product Details