Gene Transcripts

Gene Symbol
Cxcl15
Entrez Gene ID
20309
Gene Name
chemokine (C-X-C motif) ligand 15
Gene Aliases
Il8, Scyb15, lungkine, weche
Chromosome Location
Chr. 5: 90794534 - 90803067 on Build GRCm38
UniGene ID
Mm.64326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011339.2 NP_035469.1 4 345
GenBank mRNA AF082859.1 - 4 308
AK142955.1 - 4 347
BC061138.1 - 4 338 30298837

View Full Product Details