Gene Transcripts

Gene Symbol
Six2
Entrez Gene ID
20472
Gene Name
sine oculis-related homeobox 2
Gene Aliases
Chromosome Location
Chr. 17: 85662735 - 85688254 on Build GRCm38
UniGene ID
Mm.5039
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011380.2 NP_035510.1 2 1724
GenBank mRNA AK142139.1 - 1 541
D83147.1 - 2 1713

View Full Product Details