Gene Transcripts

Gene Symbol
Neto1
Entrez Gene ID
246317
Gene Name
neuropilin (NRP) and tolloid (TLL)-like 1
Gene Aliases
AI851453, Btcl1, C130005O10Rik
Chromosome Location
Chr. 18: 86394952 - 86506721 on Build GRCm38
UniGene ID
Mm.329578
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144946.4 NP_659195.3 4 1387
GenBank mRNA AF448840.1 - 4 720
AK032510.1 - 2 255
AK043710.1 - 4 723
AK081325.1 - 4 1388
AY138990.1 - 4 456
BC051145.1 - 4 747 6590345

View Full Product Details