Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146059.1 | NP_001139531.1 | 16 | 1775 | |
NM_001146060.1 | NP_001139532.1 | 16 | 1855 | ||
NM_146228.4 | NP_666340.3 | 16 | 1772 | ||
GenBank mRNA | AB107016.1 | - | 16 | 1772 | |
AF481142.1 | - | 15 | 1668 | ||
AK030815.1 | - | 9 | 909 | ||
AK053076.1 | - | 16 | 1745 | ||
AK131149.1 | - | 16 | 1774 | ||
AK155451.1 | - | 16 | 1841 | ||
BC117928.1 | - | 15 | 1725 | 40097635 | |
BC117929.1 | - | 15 | 1722 | 40097636 |