Gene Transcripts

Gene Symbol
Mab21l3
Entrez Gene ID
242125
Gene Name
mab-21-like 3 (C. elegans)
Gene Aliases
Chromosome Location
Chr. 3: 101813076 - 101836286 on Build GRCm38
UniGene ID
Mm.32900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172295.4 NP_758499.3 2 409
GenBank mRNA AK029040.1 - 2 148
AK161057.1 - 2 326
AK166928.1 - 159
AK172389.1 - 2 358
BC037703.1 - 2 346 4166596
CN663841.1 - 2 409

View Full Product Details