Gene Transcripts

Gene Symbol
Muc15
Entrez Gene ID
269328
Gene Name
mucin 15
Gene Aliases
4732460E09, D730046L02Rik, MUC-15
Chromosome Location
Chr. 2: 110721494 - 110739537 on Build GRCm38
UniGene ID
Mm.32566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290786.1 NP_001277715.1 2 522
NM_172979.3 NP_766567.1 2 522
GenBank mRNA AK052840.1 - 2 409
AK142590.1 - 2 409
AY198336.1 - 2 438
AY198337.1 - 2 405
BC094506.1 - 2 405 5341608
BC116796.1 - 2 401 40126115
BC119176.1 - 2 401 8733925

View Full Product Details