Gene Transcripts

Gene Symbol
Ppm1k
Entrez Gene ID
243382
Gene Name
protein phosphatase 1K (PP2C domain containing)
Gene Aliases
2900063A19Rik, A930026L03Rik, PP2Cm
Chromosome Location
Chr. 6: 57506502 - 57535516 on Build GRCm38
UniGene ID
Mm.396893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175523.4 NP_780732.1 2 468
GenBank mRNA AK044610.1 - 2 538
BC092238.1 - 2 468 30642218
BC138473.1 - 1 274 8861493
BC138474.1 - 1 274 8861494

View Full Product Details