Gene Transcripts

Gene Symbol
Plch2
Entrez Gene ID
269615
Gene Name
phospholipase C, eta 2
Gene Aliases
A930027K05Rik, PLCeta2, Plc-eta2, Plcl4
Chromosome Location
Chr. 4: 154983115 - 155064091 on Build GRCm38
UniGene ID
Mm.379458
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113360.2 NP_001106831.1 5 979
NM_175556.4 NP_780765.2 5 979
GenBank mRNA AK044619.1 - 5 978
AK049970.1 - 5 823
AK078731.1 - 5 750
AK129147.1 - 5 679
AY966876.1 - 5 757
DQ176851.1 - 5 979

View Full Product Details