Gene Transcripts

Gene Symbol
Npr1
Entrez Gene ID
18160
Gene Name
natriuretic peptide receptor 1
Gene Aliases
AI893888, GC-A, NPR-A, NPRA, Pndr
Chromosome Location
Chr. 3: 90450592 - 90465928 on Build GRCm38
UniGene ID
Mm.4627
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008727.5 NP_032753.5 5, 6 1621
GenBank mRNA BC110659.1 - 5, 6 1501
BC139767.1 - 5, 6 1480
J05504.1 - 6, 7 1605
L31932.1 - 5, 6 1269

View Full Product Details